accessibility menu, dialog, popup

UserName

Viewing profile as user:
NFF

CAS RN 34941-90-7

IUPAC Name: 2,4-difluoropyridine
Synonyms: 2,4-difluoropyridine
Molecular Weight (g/mol): 115.08
Molecular Formula: C5H3F2N
InChi Key: WLAKUAILRGATSL-UHFFFAOYSA-N
SMILES: FC1=CC(F)=NC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results