CAS RN 349477-49-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Thermo Scientific™ High-Purity Cyclohexyl Maltoside Detergents
Cyclohexyl maltoside detergents are a class of specialized non-ionic detergents designed for the solubilization, stabilization, and manipulation of biomolecules, particularly membrane proteins.