accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHNSNH2H2N

CAS RN 34954-13-7

IUPAC Name: 5-amino-1H-pyrazole-4-carbothioamide
Synonyms: 3-amino-2H-pyrazole-4-carbothioamide
Molecular Weight (g/mol): 142.18
Molecular Formula: C4H6N4S
InChi Key: VRWNWVXHINBCSC-UHFFFAOYSA-N
SMILES: NC(=S)C1=C(N)NN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results