accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 34957-73-8

IUPAC Name: methyl 9-hydroxynonanoate
Synonyms: methyl 9-hydroxynonanoate
Molecular Weight (g/mol): 188.27
Molecular Formula: C10H20O3
InChi Key: RIZOOQYPYGPBOC-UHFFFAOYSA-N
SMILES: COC(=O)CCCCCCCCO
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results