accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 34985-37-0

IUPAC Name: 2-methylbutan-1-amine
Synonyms: 2-methylbutan-1-amine
Molecular Weight (g/mol): 87.17
Molecular Formula: C5H13N
InChi Key: VJROPLWGFCORRM-UHFFFAOYNA-N
SMILES: CCC(C)CN
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results