accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 34985-41-6

IUPAC Name: 7-methoxy-2,3-dihydro-1H-inden-1-one
Synonyms: 7-methoxy-2,3-dihydroinden-1-one
Molecular Weight (g/mol): 162.19
Molecular Formula: C10H10O2
InChi Key: CZXBVBATQPHSSL-UHFFFAOYSA-N
SMILES: COC1=CC=CC2=C1C(=O)CC2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results