accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 34993-51-6

O

CAS RN 34993-51-6

IUPAC Name: 3-phenylcyclohexan-1-one
Synonyms: 3-phenylcyclohexan-1-one
Molecular Weight (g/mol): 174.24
Molecular Formula: C12H14O
InChi Key: CJAUDSQXFVZPTO-UHFFFAOYNA-N
SMILES: O=C1CCCC(C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)