accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHFOO

CAS RN 350-29-8

IUPAC Name: 3-fluoro-4-hydroxybenzoate
Synonyms: 3-fluoro-4-hydroxybenzoate
Molecular Weight (g/mol): 155.11
Molecular Formula: C7H4FO3
InChi Key: IUSDEKNMCOUBEE-UHFFFAOYSA-M
SMILES: OC1=CC=C(C=C1F)C([O-])=O
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)