accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 350-30-1

IUPAC Name: 2-chloro-1-fluoro-4-nitrobenzene
Synonyms: 2-chloro-1-fluoro-4-nitrobenzene
Molecular Weight (g/mol): 175.54
Molecular Formula: C6H3ClFNO2
InChi Key: DPHCXXYPSYMICK-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(F)C(Cl)=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)