accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOF(Z)

CAS RN 350-90-3

IUPAC Name: (2Z)-2-fluoro-3-phenylprop-2-enoic acid
Synonyms: cinnamic acid, α-fluoro-
Molecular Weight (g/mol): 166.15
Molecular Formula: C9H7FO2
InChi Key: QONCEXMULRJPPY-VURMDHGXSA-N
SMILES: OC(=O)C(\F)=C\C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)