accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFO

CAS RN 350-92-5

IUPAC Name: 1,1,1-trifluoro-3-phenylpropan-2-one
Synonyms: 1,1,1-trifluoro-3-phenylpropan-2-one
Molecular Weight (g/mol): 188.15
Molecular Formula: C9H7F3O
InChi Key: IAJKTOIWQHTZOS-UHFFFAOYSA-N
SMILES: FC(F)(F)C(=O)CC1=CC=CC=C1
Boiling Point 
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results