accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 35034-22-1

IUPAC Name: 2-methyl-1H-imidazole-5-carbaldehyde
Synonyms: 2-methyl-3H-imidazole-4-carbaldehyde
Molecular Weight (g/mol): 110.12
Molecular Formula: C5H6N2O
InChi Key: ZWULFIBGPXWGFG-UHFFFAOYSA-N
SMILES: CC1=NC=C(N1)C=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results