accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 350493-08-2

IUPAC Name: 1-butyl-2,3-dimethyl-1H-imidazol-3-ium; trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane
Synonyms: 3-butyl-1,2-dimethylimidazol-1-ium; bistriflylimide anion
Molecular Weight (g/mol): 433.38
Molecular Formula: C11H17F6N3O4S2
InChi Key: UCCKRVYTJPMHRO-UHFFFAOYSA-N
SMILES: CCCCN1C=C[N+](C)=C1C.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results