accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHN(R)OHO(R)OH(S)O(S)O(R)OH(S)OH(R)HO(R)HNH3CO(R)HO

CAS RN 35061-50-8

IUPAC Name: N-[(3R,4R,5S,6R)-5-{[(2S,3R,4R,5S,6R)-3-acetamido-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-2,4-dihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide
Synonyms: N,N-diacetylchitobiose N,N'-diacetylchitobioside N,N'-diacetylchitobiose
Molecular Weight (g/mol): 424.4
Molecular Formula: C16H28N2O11
InChi Key: CDOJPCSDOXYJJF-CBTAGEKQSA-N
SMILES: CC(=O)N[C@H]1C(O)O[C@H](CO)[C@@H](O[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2NC(C)=O)[C@@H]1O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results