accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3507-41-3

IUPAC Name: 5-chloro-2-(methylsulfanyl)-1,3-benzothiazole
Synonyms: 5-chloro-2-(methylsulfanyl)-1,3-benzothiazole
Molecular Weight (g/mol): 215.71
Molecular Formula: C8H6ClNS2
InChi Key: YINRVZUJQJFNES-UHFFFAOYSA-N
SMILES: CSC1=NC2=CC(Cl)=CC=C2S1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results