accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CBr

CAS RN 3508-00-7

IUPAC Name: 1-bromoheptadecane
Synonyms: 1-bromoheptadecane
Molecular Weight (g/mol): 319.37
Molecular Formula: C17H35Br
InChi Key: HHSDZLLPIXMEIU-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCBr
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Boiling Point 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)