accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOCH3OH3CO

CAS RN 35086-59-0

IUPAC Name: 3-acetyl-5-(acetyloxy)phenyl acetate
Synonyms: 3-acetyl-5-(acetyloxy)phenyl acetate
Molecular Weight (g/mol): 236.22
Molecular Formula: C12H12O5
InChi Key: QODJHYBESCIPOG-UHFFFAOYSA-N
SMILES: CC(=O)OC1=CC(=CC(OC(C)=O)=C1)C(C)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results