accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COClF

CAS RN 351-52-0

IUPAC Name: 4-(chloromethyl)-2-fluoro-1-methoxybenzene
Synonyms: 4-(chloromethyl)-2-fluoro-1-methoxybenzene
Molecular Weight (g/mol): 174.6
Molecular Formula: C8H8ClFO
InChi Key: DDAXEANMRGIVDY-UHFFFAOYSA-N
SMILES: COC1=CC=C(CCl)C=C1F
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)