CAS RN 351-87-1
IUPAC Name:
3-oxo-N-[4-(trifluoromethyl)phenyl]butanamide
Synonyms:
3-oxo-N-[4-(trifluoromethyl)phenyl]butanamide
Molecular Weight (g/mol):
245.2
Molecular Formula:
C11H10F3NO2
InChi Key:
GGDAWUSJZZHVBR-UHFFFAOYSA-N
SMILES:
CC(=O)CC(=O)NC1=CC=C(C=C1)C(F)(F)F
1
–
1
of
1
results
1
3-Oxo-N-(4-trifluoromethylphenyl)butyramide 98.0+%, TCI America™
CAS: 351-87-1 Molecular Formula: C11H10F3NO2 Molecular Weight (g/mol): 245.201 MDL Number: MFCD00231301 InChI Key: GGDAWUSJZZHVBR-UHFFFAOYSA-N Synonym: 4′C-(Trifluoromethyl)acetoacetanilide PubChem CID: 1472078 IUPAC Name: 3-oxo-N-[4-(trifluoromethyl)phenyl]butanamide SMILES: CC(=O)CC(=O)NC1=CC=C(C=C1)C(F)(F)F