CAS RN 351003-21-9
CAS RN 351003-21-9
IUPAC Name:
2-bromo-4-fluoro-1-(trifluoromethyl)benzene
Synonyms:
2-bromo-4-fluoro-1-(trifluoromethyl)benzene
Molecular Weight (g/mol):
243
Molecular Formula:
C7H3BrF4
InChi Key:
XBBGYSIEJHXPLL-UHFFFAOYSA-N
SMILES:
FC1=CC(Br)=C(C=C1)C(F)(F)F
1
–
1
of
1
results
1
2-Bromo-4-fluorobenzotrifluoride 98.0+%, TCI America™

CAS: 351003-21-9 Molecular Formula: C7H3BrF4 Molecular Weight (g/mol): 243.00 MDL Number: MFCD03094041 InChI Key: XBBGYSIEJHXPLL-UHFFFAOYSA-N Synonym: 2-bromo-4-fluorobenzotrifluoride,2-bromo-4-fluoro-1-trifluoromethyl benzene,1-bromo-2-trifluoromethyl-5-fluorobenzene,benzene, 2-bromo-4-fluoro-1-trifluoromethyl,2-bromo-4-florobenzotrifluoride,2-bromo-4-fluoro-1-trifluoromethyl-benzene,pubchem1636,acmc-209icx,ksc911k1l,2-bromo-4-fluorobenzo trifluoride PubChem CID: 2734909 IUPAC Name: 2-bromo-4-fluoro-1-(trifluoromethyl)benzene SMILES: FC1=CC(Br)=C(C=C1)C(F)(F)F