accessibility menu, dialog, popup

UserName

Viewing profile as user:
SOOClNF

CAS RN 351003-23-1

IUPAC Name: 3-cyano-4-fluorobenzene-1-sulfonyl chloride
Synonyms: 3-cyano-4-fluorobenzenesulfonyl chloride
Molecular Weight (g/mol): 219.61
Molecular Formula: C7H3ClFNO2S
InChi Key: KDVZADPGGPBAAK-UHFFFAOYSA-N
SMILES: FC1=C(C=C(C=C1)S(Cl)(=O)=O)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results