accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 351003-52-6

IUPAC Name: 4-bromo-2-chlorobenzene-1-sulfonyl chloride
Synonyms: 4-bromo-2-chlorobenzenesulfonyl chloride
Molecular Weight (g/mol): 289.95
Molecular Formula: C6H3BrCl2O2S
InChi Key: OECYEHWJPRPCOH-UHFFFAOYSA-N
SMILES: ClC1=CC(Br)=CC=C1S(Cl)(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)