accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 35103-79-8

IUPAC Name: caesium(1+) hydrate hydroxide
Synonyms: caesium(1+) hydrate hydroxide
Molecular Weight (g/mol): 167.93
Molecular Formula: CsH3O2
InChi Key: ABSOMGPQFXJESQ-UHFFFAOYSA-M
SMILES: O.[OH-].[Cs+]
Percent Purity 
  • (2)
  • (4)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (6)
  • (2)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)