accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3512-80-9

IUPAC Name: 2,6-dimethylpyridin-4-amine
Synonyms: 2,6-dimethyl-pyridin-4-ylamine
Molecular Weight (g/mol): 122.17
Molecular Formula: C7H10N2
InChi Key: ZJXMKPARTVOUAM-UHFFFAOYSA-N
SMILES: CC1=CC(N)=CC(C)=N1
Molecular Weight (g/mol) 
  • (1)
  • (5)
Quantity 
  • (3)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (5)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)