accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHH2CHO

CAS RN 3513-81-3

IUPAC Name: 2-methylidenepropane-1,3-diol
Synonyms: 2-methylidenepropane-1,3-diol
Molecular Weight (g/mol): 88.11
Molecular Formula: C4H8O2
InChi Key: JFFYKITVXPZLQS-UHFFFAOYSA-N
SMILES: OCC(=C)CO
Boiling Point 
  • (1)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)