accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 351422-73-6

IUPAC Name: (3-carbamoylphenyl)boronic acid
Synonyms: 3-carbamoylphenylboronic acid
Molecular Weight (g/mol): 164.96
Molecular Formula: C7H8BNO3
InChi Key: WDGWHKRJEBENCE-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC=CC(=C1)B(O)O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results