accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 35144-22-0

IUPAC Name: 2-methanesulfonyl-4,6-dimethylpyrimidine
Synonyms: 2-methanesulfonyl-4,6-dimethylpyrimidine
Molecular Weight (g/mol): 186.23
Molecular Formula: C7H10N2O2S
InChi Key: ZHPSNGCLCHWTRG-UHFFFAOYSA-N
SMILES: CC1=CC(C)=NC(=N1)S(C)(=O)=O
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)