accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 35150-07-3

IUPAC Name: tert-butyl 2-carbamoylpyrrolidine-1-carboxylate
Synonyms: tert-butyl 2-carbamoylpyrrolidine-1-carboxylate
Molecular Weight (g/mol): 214.27
Molecular Formula: C10H18N2O3
InChi Key: PITJAAIPVBVRAO-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1CCCC1C(N)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results