accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3516-95-8

IUPAC Name: 1-(2-hydroxyphenyl)-3-phenylpropan-1-one
Synonyms: 1-(2-hydroxyphenyl)-3-phenylpropan-1-one
Molecular Weight (g/mol): 226.28
Molecular Formula: C15H14O2
InChi Key: JCPGMXJLFWGRMZ-UHFFFAOYSA-N
SMILES: OC1=CC=CC=C1C(=O)CCC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results