accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 35161-71-8

IUPAC Name: methyl(prop-2-yn-1-yl)amine
Synonyms: 2-propyn-1-amine, N-methyl-
Molecular Weight (g/mol): 69.11
Molecular Formula: C4H7N
InChi Key: HQFYIDOMCULPIW-UHFFFAOYSA-N
SMILES: CNCC#C
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)