accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 352-11-4

IUPAC Name: 1-(chloromethyl)-4-fluorobenzene
Synonyms: P-fluorobenzyl chloride
Molecular Weight (g/mol): 144.57
Molecular Formula: C7H6ClF
InChi Key: IZXWCDITFDNEBY-UHFFFAOYSA-N
SMILES: FC1=CC=C(CCl)C=C1
Boiling Point 
  • (2)
  • (5)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (5)
Quantity 
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (5)
  • (2)
  • (2)
  • (2)
  • (5)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)