accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaH3CNH3CONCH3CH3CH3H3CSOOOSOOO(?)

CAS RN 3520-42-1

IUPAC Name: sodium 3,6-bis(diethylamino)-9-(2,4-disulfonatophenyl)-2,7-dimethyl-10λ⁴-xanthen-10-ylium
Synonyms: sulforhodamine B acid rhodamine B amido rhodamine B xylene red B acid red XB acid red 52 food red 106 kiton rhodamine B lissamine rhodamine
Molecular Weight (g/mol): 608.7
Molecular Formula: C29H33N2NaO7S2
InChi Key: XJENLUNLXRJLEZ-UHFFFAOYSA-M
SMILES: [Na+].CCN(CC)C1=CC2=[O+]C3=CC(N(CC)CC)=C(C)C=C3C(C3=CC=C(C=C3S([O-])(=O)=O)S([O-])(=O)=O)=C2C=C1C
Quantity 
  • (1)
  • (4)
  • (2)
Color 
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (3)
  • (1)
  • (1)