accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 352018-88-3

IUPAC Name: (5-phenyl-1,3-oxazol-4-yl)methanol
Synonyms: (5-phenyl-1,3-oxazol-4-yl)methanol
Molecular Weight (g/mol): 175.19
Molecular Formula: C10H9NO2
InChi Key: LVHWTAMRDRDXJP-UHFFFAOYSA-N
SMILES: OCC1=C(OC=N1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results