accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 352018-95-2

IUPAC Name: 4-iodo-2,1,3-benzothiadiazole
Synonyms: 4-iodo-2,1,3-benzothiadiazole
Molecular Weight (g/mol): 262.07
Molecular Formula: C6H3IN2S
InChi Key: ROPRWVSWECJQIC-UHFFFAOYSA-N
SMILES: IC1=CC=CC2=NSN=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results