accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 352018-96-3

IUPAC Name: 1-(4-isothiocyanatophenyl)-1H-pyrazole
Synonyms: 1-(4-isothiocyanatophenyl)pyrazole
Molecular Weight (g/mol): 201.25
Molecular Formula: C10H7N3S
InChi Key: GLAOJNMWLIFKCG-UHFFFAOYSA-N
SMILES: S=C=NC1=CC=C(C=C1)N1C=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results