accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 35203-44-2

IUPAC Name: 1-propyl-1H-imidazole
Synonyms: 1-propylimidazole
Molecular Weight (g/mol): 110.16
Molecular Formula: C6H10N2
InChi Key: IYVYLVCVXXCYRI-UHFFFAOYSA-N
SMILES: CCCN1C=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results