accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOOI

CAS RN 35204-44-5

IUPAC Name: ethyl 6-iodo-4-oxo-4H-chromene-2-carboxylate
Synonyms: ethyl 6-iodo-4-oxochromene-2-carboxylate
Molecular Weight (g/mol): 344.1
Molecular Formula: C12H9IO4
InChi Key: HUJFMOAXSPSGDU-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC(=O)C2=CC(I)=CC=C2O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results