accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 35216-11-6

IUPAC Name: tetradec-7-yne
Synonyms: tetradec-7-yne
Molecular Weight (g/mol): 194.36
Molecular Formula: C14H26
InChi Key: AFNWSIIBAYUTTL-UHFFFAOYSA-N
SMILES: CCCCCCC#CCCCCCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results