CAS RN 352341-26-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Luciferase Inhibitor I
The Luciferase Inhibitor I, also referenced under CAS 352341-26-5, controls the biological activity of Luciferase.