accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3524-32-1

IUPAC Name: 1,3-dimethyl-1H-pyrazol-5-amine
Synonyms: 2,5-dimethylpyrazol-3-amine
Molecular Weight (g/mol): 111.15
Molecular Formula: C5H9N3
InChi Key: ZFDGMMZLXSFNFU-UHFFFAOYSA-N
SMILES: CN1N=C(C)C=C1N
Molecular Weight (g/mol) 
  • (5)
  • (1)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (5)
  • (5)
  • (1)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)