accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3524-73-0

IUPAC Name: 5-methylhex-1-ene
Synonyms: 1-hexene, 5-methyl-
Molecular Weight (g/mol): 98.19
Molecular Formula: C7H14
InChi Key: JIUFYGIESXPUPL-UHFFFAOYSA-N
SMILES: CC(C)CCC=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results