accessibility menu, dialog, popup

UserName

Viewing profile as user:
BSHHOOH

CAS RN 352526-01-3

IUPAC Name: (3-sulfanylphenyl)boronic acid
Synonyms: 3-sulfanylphenylboronic acid
Molecular Weight (g/mol): 153.99
Molecular Formula: C6H7BO2S
InChi Key: OBCKMTLUSKYKIW-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(S)=CC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)