accessibility menu, dialog, popup

UserName

Viewing profile as user:
CClClZnI

CAS RN 352530-43-9

IUPAC Name: 2,5-dichlorobenzen-1-ide; iodozincylium
Synonyms: 2,5-dichlorobenzen-1-ide; iodozincylium
Molecular Weight (g/mol): 338.27
Molecular Formula: C6H3Cl2IZn
InChi Key: ZNRGDLRHRVNWNM-UHFFFAOYSA-M
SMILES: [Zn+]I.ClC1=CC=C(Cl)[C-]=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results