accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 352535-83-2

IUPAC Name: (5-chloro-2-fluorophenyl)boronic acid
Synonyms: 5-chloro-2-fluorophenylboronic acid
Molecular Weight (g/mol): 174.36
Molecular Formula: C6H5BClFO2
InChi Key: GGTUVWGMCFXUAS-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(Cl)=CC=C1F
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results