accessibility menu, dialog, popup

UserName

Viewing profile as user:
BBrFHOOH

CAS RN 352535-97-8

IUPAC Name: (3-bromo-2-fluorophenyl)boronic acid
Synonyms: 3-bromo-2-fluorophenylboronic acid
Molecular Weight (g/mol): 218.82
Molecular Formula: C6H5BBrFO2
InChi Key: QCLOSORNRRMFIA-UHFFFAOYSA-N
SMILES: OB(O)C1=C(F)C(Br)=CC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results