accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 352637-44-6

IUPAC Name: 6-bromopyrazolo[1,5-a]pyrimidine-3-carbonitrile
Synonyms: 6-bromopyrazolo[1,5-a]pyrimidine-3-carbonitrile
Molecular Weight (g/mol): 223.03
Molecular Formula: C7H3BrN4
InChi Key: IDYCEZBYNPADDN-UHFFFAOYSA-N
SMILES: BrC1=CN2N=CC(C#N)=C2N=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results