accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 353-54-8

IUPAC Name: tribromo(fluoro)methane
Synonyms: tribromofluoromethane
Molecular Weight (g/mol): 270.72
Molecular Formula: CBr3F
InChi Key: IHZAEIHJPNTART-UHFFFAOYSA-N
SMILES: FC(Br)(Br)Br
Boiling Point 
  • (3)
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (1)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (4)
  • (1)
  • (5)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)