accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 35302-72-8

IUPAC Name: 2,2,2-trichloro-1-(1H-pyrrol-2-yl)ethan-1-one
Synonyms: 2,2,2-trichloro-1-(1H-pyrrol-2-yl)ethanone
Molecular Weight (g/mol): 212.45
Molecular Formula: C6H4Cl3NO
InChi Key: BBFDGMDENAEMKF-UHFFFAOYSA-N
SMILES: ClC(Cl)(Cl)C(=O)C1=CC=CN1
Molecular Weight (g/mol) 
  • (4)
  • (1)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)