accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 353258-31-8

IUPAC Name: ethyl 8-chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
Synonyms: ethyl 8-chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
Molecular Weight (g/mol): 292.64
Molecular Formula: C11H8ClF3N2O2
InChi Key: AKACORAIGDTIFR-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CN2C=C(C=C(Cl)C2=N1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results