accessibility menu, dialog, popup

UserName

Viewing profile as user:
SNBr

CAS RN 35342-50-8

IUPAC Name: 5-bromo-2,4-diphenyl-1,3-thiazole
Synonyms: 5-bromo-2,4-diphenyl-1,3-thiazole
Molecular Weight (g/mol): 316.22
Molecular Formula: C15H10BrNS
InChi Key: KKDPGKYZSJFNLZ-UHFFFAOYSA-N
SMILES: BrC1=C(N=C(S1)C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results